Safety cabinet type 90 UTS ergo line S with classical door technology. For the storage of flammable liquids. Type-tested by MPA/ TÜV SÜD according to DIN EN 14470-1, laboratory furniture standard DIN EN 14727, EK5/AK4 09-10:2009 and TRGS 510 Annex 3 1 * 1 items
Supplier: DUPERTHAL
Safety cabinet type 90 UTS ergo line S with classical door technology. For the storage of flammable liquids. Type-tested by MPA/ TÜV SÜD according to DIN EN 14470-1, laboratory furniture standard DIN EN 14727, EK5/AK4 09-10:2009 and TRGS 510 Annex 3 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.