Microtest manifolds
Supplier: DRUMMOND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3-000-093 3-000-094 3-000-090 3-000-096 3-000-097 3-000-098 3-000-101 3-000-102 3-000-103 3-000-104 3-000-099
DRUM3-000-093EA 672 GBP
DRUM3-000-093 DRUM3-000-094 DRUM3-000-090 DRUM3-000-096 DRUM3-000-097 DRUM3-000-098 DRUM3-000-101 DRUM3-000-102 DRUM3-000-103 DRUM3-000-104 DRUM3-000-099
Microtest manifolds
Manifolds
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection