Calibration gases
Supplier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6810687 6811647 6811130 6811131 6810391 6810394 3713477 6810387 6812106
DRAE6810687EA 217 GBP
DRAE6810687 DRAE6811647 DRAE6811130 DRAE6811131 DRAE6810391 DRAE6810394 DRAE3713477 DRAE6810387 DRAE6812106
Calibration gases
Standards Environmental Analysis Standards
Calibration gas with nitrogen background in non refillable cylinder.
Ordering information: Requires a regulator and a delivery tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable