Anti-8-Hydroxyguanosine Goat Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

DR1001-100
DR1001-100EA 447 GBP
DR1001-100
Anti-8-Hydroxyguanosine Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR