MOLYKOTE® 111 Compound Valve Lubricant and Sealant
Supplier: DOW CORNING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
23900034
DOWC23900034EA 59.2 GBP
DOWC23900034
MOLYKOTE® 111 Compound Valve Lubricant and Sealant
Lubricants
Lubrication for control and pressure plug valves, water softener and faucet valves. Sealant for vacuum and pressure systems and outdoor equipment (also shipboard) subject to washing and harsh environmental exposure, such as meters, electrical service entrance and underground connections.
  • Water resistance
  • Wide service-temperature range
  • Compatible with many plastics and elastomers
  • Low vapor pressure and volatility
Damping medium for dash pots in electrical and electronic equipment. As an anti-stick and a sealant for transformer gasket and equipmentMore Product Information