Disinfectant detergent, clearKlens deogen
Supplier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7522374
DIVY7522374EA 139 GBP
DIVY7522374
Disinfectant detergent, clearKlens deogen
Cleaning Agents Disinfectants
Chlorinated alkaline detergent and disinfectant for soft/medium hard water.
  • Low foaming under conditions of high turbulence
  • High active chlorine level effectively removes soils, stains and off-flavours
  • Broad spectrum disinfectant action
  • Free-rinsing, minimises water usage
Used for CIP and spray washing in pharmaceutical, cosmetic, and other industries.
Caution: Use biocides safely. Always read the label and product information before use.
Order Now
SPECIFICATIONS
Environmentally Preferable
Specific gravity 1,170 – 1,210 g/ml