Glass and stainless steel cleaner, TASKI sprint 200
Supplier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7512859
DIVY7512859EA 51.1 GBP
DIVY7512859
Glass and stainless steel cleaner, TASKI sprint 200
Cleaning Agents Detergents and Surface Cleansers
Solvent-based formulation that gives low residue and fast buff response to give streak-free finish on all surfaces including glass and stainless steel.
  • Multi-purpose for surfaces and floors
  • Suitable for use with trigger spray or bucket
  • Pleasant fragrance with ONT (Odour Neutralising Technology)