Excel wipes, Oxivir®
Supplier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100984250 100984246
DIVY100984250EA 56.1 GBP
DIVY100984250 DIVY100984246
Excel wipes, Oxivir®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Powered by AHP® technology. Oxivir® disinfectant cleaners provide an alternative to traditional disinfectants by delivering fast acting, broad spectrum disinfection with enhanced cleaning power that is staff and environmentally friendly.
  • Broad efficacy
  • Short contact times keep the surface wet, thus ensuring disinfectant efficacy
  • Excellent cleaning performance
  • Positive user and surface safety profile
  • Fragrance-free
  • Active breaks down into water and oxygen
  • No rinsing required
Oxivir® Excel wipe CE is a one-step cleaner disinfectant wipe with broadMore Product Information