Velostat® Round Container
Supplier: DESCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
57016
DESC57016EA 4 GBP
DESC57016
Velostat® Round Container
Boxes Storage Boxes
Round Velostat® container ideal for static-safe storage, kitting, and in-process handling and transporting of static-sensitive assemblies and devices.
  • Black, injected molded container
  • Lid included