Surface active cleaning agent, Neutrad®
Supplier: DECON LAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NEUT5
DECNNEUT5EA 89.8 GBP
DECNNEUT5
Surface active cleaning agent, Neutrad®
Cleaning Agents Detergents and Surface Cleansers
A surface active cleaning agent and/or decontaminant, for use on materials which may be corroded, etched, discoloured or weakened by acidic or alkaline cleaning agents. Ideal for cleaning all non ferrous metals such as aluminium, zinc, copper, silver and brass.
  • Phosphate- and enzyme-free
  • Non toxic
  • Neutral pH 7
OECD 301E (ISO method 7827-1986 (E))