Anti-IgG Peroxidase Conjugate Goat Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

DC01L-200
DC01L-200EA 208 GBP
DC01L-200
Anti-IgG Peroxidase Conjugate Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IgG Peroxidase Conjugate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR