Sharps Containers, SHARPSGUARD® eco yellow
Supplier: DANIELS HEALTHCARE LIMITED
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DD442NRYLR DD442YLR DD472YLR DD474YLR DD475YLR DD476YLR DD477YLR DD477WYLR DD509YLR DD440YLR DD439YLR DD471YLR
DANIDD442NRYLREA 170 GBP
DANIDD442NRYLR DANIDD442YLR DANIDD472YLR DANIDD474YLR DANIDD475YLR DANIDD476YLR DANIDD477YLR DANIDD477WYLR DANIDD509YLR DANIDD440YLR DANIDD439YLR DANIDD471YLR
Sharps Containers, SHARPSGUARD® eco yellow
Bins Sharps Containers
The SHARPSGUARD® eco yellow range is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic or cytostatic.
  • Sustainable alternative made from at least 40% recycled plastic
  • Small models can be easily fitted into a bag or glove compartment
  • Models eco yellow 7 and above are supplied with a robust pail handle for easy transportation
These containers are for disposal by incineration or other authorised process and designed for the consignment of waste categorised by the following EWC codes: 18 01 01, 18 01 03*, 18 01 09More Product Information