Cut resistant gloves, Chainexpert Wrist
Supplier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2533001R0302 2533002R0302 2533004R0302 2533005R0302 2533006R0302
DALL2533001R0302EA 285 GBP
DALL2533001R0302 DALL2533002R0302 DALL2533004R0302 DALL2533005R0302 DALL2533006R0302
Cut resistant gloves, Chainexpert Wrist
Gloves
These chainmail wrist gloves with self adjusting wrist spring are manufactured from stainless steel. The use of steel with a shape retention memory ensures perfect resistance to stretching over a long period for ultimate protection.
  • Steel spring strap adapts itself to any wrist shape due to it’s natural elasticity
  • Bacteria proof, easy to clean for complete hygiene
  • Self adjustable, molding itself to the hand without need for any other type of closure
  • Promotes 100% wear time with no straps or press stud fastenings
  • No lateral slit on glove completely
More Product Information