Sealable Standup Pouches
Supplier: DAKLAPACK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
481121 481125
DAKL481121EA 126 GBP
DAKL481121 DAKL481125
Sealable Standup Pouches
Bags Storage Bags
These pouches are intended for use as a one time use pouch for food and even non food products. The pouch is designed with an opening bottom gusset which means the pouch will stand up on its own.
  • Available in many different sizes
  • High quality laminated foil offers high barrier properties
  • With a round bottom gusset and an opening without zip
Solids, liquids or powders can all be store in these pouches and easily sealed closed using a heat sealer.