Bicinchoninic acid disodium salt hydrate

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D8284-10G D8284-1G D8284-25G D8284-5G
D8284-10GEA 475 GBP
D8284-10G D8284-1G D8284-25G D8284-5G
Bicinchoninic acid disodium salt hydrate
Assays Protein Assays

Bicinchoninic acid is used for the determination of protein concentration. It is responsible for the making of intense purple complex with cuprous ion (Cu+) in the presence of alkaline conditions. The amount of Cu+ is in relation with the protein concentration.


  • Assay: ≥98%
  • Form: Powder
  • Colour: White to light yellow


Bicinchoninic acid disodium salt hydrate has been used to monitor the levels of reducing sugars, thereby helping in the detection of chitinases. It has also been used to determine the amount of cellulose reducing ends (micromoles/g cellulose).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0865773249c488075d9c842c176468842a143c69&vid=fc54a7e38c73b2fa386ec2cbbb91a13a8638d791&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR