J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D014-00EA 29.7 GBP
D014-00 D014-04 D012-00 D013-00 D013-02 D013-04 D014-02 D015-00 D015-02 D016-00 D016-02 D016-04
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Buffers Biological Buffers
DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities