EPA screw neck vials, Chromacol™
Supplier: CHROMACOL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40-EPAVCS 20-EPAVCSA 20-EPAVCS-PC 40-EPAVCS-PC 60-EPAVCS
CRMA40-EPAVCSEA 0 GBP
CRMA40-EPAVCS CRMA20-EPAVCSA CRMA20-EPAVCS-PC CRMA40-EPAVCS-PC CRMA60-EPAVCS
EPA screw neck vials, Chromacol™
Vials
Level 100 EPA vials: These are processed and packaged under a strict registered ISO Quality Management System in the same manner as Level 300 products. Level 100 products are not certified or pre-cleaned. Every case of product is labelled with its production number and is custody sealed to ensure reliable chain-of-custody. These convenience kits are recommended for discrete water sampling under EPA 40 CFR 136 'Guidelines for Establishing Test Procedures for the Analysis of Pollutants" and EPA 40 CFR 141 "National Interim Primary Drinking Water Regulations: Control of Trihalomethanes inMore Product Information