Supelco® TraceCERT® Organic Reference Standard, Ametoctradin hydrochloride Metabolite M650F06
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRM18501-10MG
CRM18501-10MGEA 81.3 GBP
CRM18501-10MG
Supelco® TraceCERT® Organic Reference Standard, Ametoctradin hydrochloride Metabolite M650F06
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P264 - P270 - P301 + P312 - P501