Standard vest style fall arrest harness, Protecta®, 3M™
Supplier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1161609 1161610 1161611 1161615 1161616 1161617 1161642 1161643 1161644
CPSD1161609EA 0 GBP
CPSD1161609 CPSD1161610 115-2663 CPSD1161615 CPSD1161616 CPSD1161617 CPSD1161642 115-2664 CPSD1161644
Standard vest style fall arrest harness, Protecta®, 3M™
Fall Protection Fall Harnesses
Protecta® Standard Vest Style Fall Arrest Harness is a sleek and modern safety harness with upgraded features. It has water and stain resistant webbing which keeps it clean and dry, and protected labels that stay readable in the long term for simple and easy inspection.
  • Stain and water resistant webbing keeps the harness cleaner and dryer
  • Steel, zinc-plated black electro-deposition paint D-rings
  • Conveniently placed front D-ring helps users stay balanced and centred
  • Auto-resetting lanyard keeper provides easy parking and release of snap hooks or carabiners
More Product Information