MPX series couplings, medical grade
Supplier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MPX17839B25 MPX22639MB25 MPX22803MB25 MPX22839MB25 MPX32003 MPX32003B25 MPX32039 MPX32039B25 MPXK17839B25
CPCCMPX17839B25EA 437 GBP
CPCCMPX17839B25 CPCCMPX22639MB25 CPCCMPX22803MB25 CPCCMPX22839MB25 CPCCMPX32003 CPCCMPX32003B25 CPCCMPX32039 CPCCMPX32039B25 CPCCMPXK17839B25
MPX series couplings, medical grade
Fittings and Connectors
MPX Series couplings add ease of use and security to your most critical fluid handling applications.
  • Operating temperature: −40 to 149 °C
  • Easy to operate
  • BSE/TSE free materials
  • Parting line free hose barb
  • Ergonomic thumb latch
  • Sterilisable by autoclave, Et0, e-beam or gamma
These couplings provide optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Certifications: USP Class VI, ADCF certified and meets biocompatibility requirements.