AseptiQuik® L Connectors
Supplier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AQL17012 AQL17012HT AQL17016 AQL17016HT AQL33024 AQL33024HT
CPCCAQL17012EA 166 GBP
CPCCAQL17012 CPCCAQL17012HT 212-0823 CPCCAQL17016HT CPCCAQL33024 CPCCAQL33024HT
AseptiQuik® L Connectors
Fittings and Connectors
AseptiQuik® L connectors enable quick and easy sterile connections, in large-volume, high-flow production environments. The large-format, ³/₄", 1" hose barb and 1½" sanitary genderless design simplifies system integration and minimises the risk of operator error.
  • Genderless: Eases Single-use systems specifications with one-part number for both halves
  • Flip-Click-Pull: Intuitive three-step connection process reduces risk of operator error
  • CPC Click: Audible confirmation of connection with no additional hardware required
  • AseptiQuik PPSU enables genderless
More Product Information