Stopcocks, straight through, with tip
Supplier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
016.9202-B 016.9202.1 016.9203-B 016.9212.1
661-1181EA 14.9 GBP
661-1181 661-1183 COWI016.9203-B COWI016.9212.1
Stopcocks, straight through, with tip
Stopcocks
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance