Stoppers, flat type, PTFE
Supplier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
010.134 010.129 010.124 010.119 010.114 010.110
COWI010.134EA 19.7 GBP
COWI010.134 COWI010.129 COWI010.124 COWI010.119 COWI010.114 COWI010.110
Stoppers, flat type, PTFE
Stoppers PE/LDPE/HDPE Stoppers
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure