Stirrers tools, adjustable rotor, PTFE
Supplier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
005.608040 005.610060 005.612070 005.612090 005.708070 005.710070 005.710100 005.712100 005.712150 005.7.313.3 005.716150 005.716100 005.706070 005.7.625.6 005.6.250.15 005.7.375.3 005.7.250.3 005.616100 005.606040 005.6.625.40 005.6.500.35 005.6.375.25 005.6.313.15 005.7.500.4 005.610090
COWI005.608040EA 45.1 GBP
COWI005.608040 COWI005.610060 COWI005.612070 COWI005.612090 COWI005.708070 COWI005.710070 COWI005.710100 COWI005.712100 COWI005.712150 COWI005.7.313.3 COWI005.716150 COWI005.716100 COWI005.706070 COWI005.7.625.6 COWI005.6.250.15 COWI005.7.375.3 COWI005.7.250.3 COWI005.616100 COWI005.606040 COWI005.6.625.40 COWI005.6.500.35 COWI005.6.375.25 COWI005.6.313.15 COWI005.7.500.4 COWI005.610090
Stirrers tools, adjustable rotor, PTFE
Stirrers Stirring Tools
Adjustable rotors made of PTFE with glass filled PTFE lock nut.
  • Resistant to chemicals
  • Can be use up to 280 °C
These rotors slide over standard shafts and lock in place to give the most effective stirring patterns.