Corning® 1536-Well Microplates, Cyclic Olefin Copolymer
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4567 4570 4566
CORN4567EA 1560 GBP
CORN4567 CORN4570 CORN4566
Corning® 1536-Well Microplates, Cyclic Olefin Copolymer
Microplates HTS Microplates
Cyclic olefin copolymer (COC). Superior flatness of the plates minimises variability within a plate and between plates. The barcoded plates come with with raised well bottoms for higher sensitivity.
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Working volume is up to 10 µl
  • Flat bottom wells are compatible with automated dispensers