Storage plates and blocks, 96-well, Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3947
CORN3947EA 0 GBP
CORN3947
Storage plates and blocks, 96-well, Corning®
Microplates Deep Well Microplates
PP, non treated, without lid. Standard and large volume 96-well microplates and storage blocks to meet assay and storage requirements.
  • Choice of flat, round or V-shaped well bottom
  • Uniform skirt heights for greater robotic gripping surface
  • Resistant to many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Certified free from DNase and RNase