NAO™ Baskets for Rapid and Efficient DNA Recovery

Supplier: Copan
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3002C 3102CS01 4103CS01 30U013D01
COPA3002CEA 1020 GBP
COPA3002C COPA3102CS01 COPA4103CS01 COPA30U013D01
NAO™ Baskets for Rapid and Efficient DNA Recovery
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

NAO® Basket for rapid and efficient DNA recovery. This is a semi-permeable system designed for releasing and concentrating human DNA from swab samples or other specimens during the extraction step. Lysis in the NAO® Basket: NAO® Basket retains the lysis buffer during the lysis step of the forensic sample.



  • Application: DNA rapid recovory

  • Lyse and centrifuge in the basket: innovative product design allows the passage of DNA from samples into the tube using a tight leak-proof design that can endure vortexing, centrifugations, and various storage conditions

  • Multiple configurations: NAO® Basket are available in several formats as single pouches or bulk packages, both with 1,5 ml or 2 ml vials
  • Compatibility: NAO® Basket is compatible with commercial Nucleic Extraction Platforms
  • Forensic DNA grade: ISO 18385:2016 compliance minimises the risk of detectable human nuclear DNA contamination in products used by the global forensic community


Upon centrifugation on a high-speed centrifuge, valves in the bottom of the NAO® Basket open up and allow the sample lysate to pass through. The lysate containing the nucleic acids is efficiently recovered and collected in the provided collection tube, whereas the substrate remains in the NAO® Basket.



How to use NAO® Basket:
1. Break the FLOQSwabs® at the breaking point into the NAO® Basket.
2. Add the lysis buffer and incubate as per extraction kit instructions.
3. Upon centrifugation, the sample lysate passes through the basket and is available for testing at the bottom of the tube. Improve DNA yield from challenging samples:
The combination of NAO® Basket and FLOQSwabs® provides more than 90% DNA recovery of the reference sample collected by the swab, particularly critical for crime scene evidence testing. The 20 mm breaking point designed on the 4N6FLOQSwabs® is compatible with the NAO® Basket.



Certifications: Certification: ISO 18385:2016. The validated EO TREATMENT assesses the degradation of possible human DNA residuals. Free of amplifiable human DNA, detectable and amplifiable DNase, and RNase. Personnel STR database.


Caution: The NAO® Basket is not intended for diagnostic purposes.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR