Cleaning swabs, double-ended, CONSTIX® SC-4
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SC-4
CONTSC-4EA 585 GBP
CONTSC-4
Cleaning swabs, double-ended, CONSTIX® SC-4
Swabs Swabs for Cleaning
SC-4 consists of high quality, long staple length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles.
  • Double-ended hard, sharp points
  • High quality cotton fibres provide excellent particle entrapment and retention
  • Tightly wrapped head helps maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high quality cotton fibres provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using a methyl-cellulose binder whichMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable