QuickConnect™ Mop Handles
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CFH32 CFH70 CFH114 CFH224 CFH334
CONTCFH32EA 133 GBP
CONTCFH32 CONTCFH70 CONTCFH114 CONTCFH224 CONTCFH334
QuickConnect™ Mop Handles
Mops
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, as wellMore Product Information