Cleanroom wipes, Amplitude™ VIP-L
Supplier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AMVL0001 AMVL0002 AMVL0004
CONTAMVL0001EA 638 GBP
CONTAMVL0001 CONTAMVL0002 CONTAMVL0004
Cleanroom wipes, Amplitude™ VIP-L
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This standard weight wipe with textured surface is knitted from filament polyester. It is ideal for effective cleaning.
  • Soft textured wipe cleans surfaces without scratching
  • Extremely low in both particles and fibers
  • Good sorbency with solvents
  • Chemical resistant
The wipe is laser-cut, which seals the fibers at the edges. It is laundered and packaged in an ISO Class 4 cleanroom to further reduce contamination.