Multi channel data loggers for pH, conductivity and temperature
Supplier: CONSORT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D230 D292 D230-CH D230-UK
CONSD230EA 1940 GBP
CONSD230 CONSD292 CONSD230-CH CONSD230-UK
Multi channel data loggers for pH, conductivity and temperature
Data Loggers
Multi-parameter data-logger is a configurable large measurement channel arrays like pH/mV/Ion, conductivity, oxygen and temperature.
  • Multichannel up to 448 measurements can be performed at the same time and simultaneous displayed on the screen
  • No interference between pH/ORP/Ion and conductivity electrodes in the same solution
  • Free data acquisition software to control, collect and store data of a D230 system
  • Data-logging can start/stop automatically or at a programmable date/time
  • All data is saved in a user defined file in CSV format
  • Graphs are generated
More Product Information