Light Duty Measuring Jugs
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MEA4001G MEA4002G MEA40.5G
COLDMEA4001GEA 66.7 GBP
COLDMEA4001G COLDMEA4002G COLDMEA40.5G
Light Duty Measuring Jugs
Beakers
Stainless steel measuring jugs meet international pharma quality compliance regulations.
  • All sizes available as standard, off-the-shelf for delivery within days
  • 304 pharmaceutical grade stainless steel
  • Graduated in pints and litres
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Material Grade, Surface Finish, and TSE certification available upon request
Made from 304 grade stainless steel, these measuring jugs are tapered and come with either an open or closed handle. They feature crevice-free and pit-freeMore Product Information