Scoops
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CFS6001P CFS6002P
COLDCFS6001PEA 623 GBP
COLDCFS6001P COLDCFS6002P
Scoops
Scoops
Stainless steel 316L.
  • Heavy-duty
  • Crevice- and pit-free
  • Polished throughout
  • With round handle