Beakers, low form, stainless steel
Supplier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BEA4040C BEA4070C BEA4125C BEA4250C
COLDBEA4040CEA 156 GBP
COLDBEA4040C COLDBEA4070C COLDBEA4125C COLDBEA4250C
Beakers, low form, stainless steel
Beakers
Stainless steel 304 grade.
  • Made in one piece
  • Crevice and pit free
  • Polished throughout
Ideal for corrosive materials. Robust and durable. With spout.