1H-Pyrazole-4-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PN-8851-5G PN-8851-1G
COBBPN-8851-5GEA 0 GBP
COBBPN-8851-5G COBBPN-8851-1G
1H-Pyrazole-4-boronic acid pinacol ester
1H-Pyrazole-4-boronic acid pinacol ester
Formula: C₉H₁₅BN₂O₂
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR