2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: tert-Butyl 4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)piperazine-1-carboxylate

PN-8722-5G PN-8722-1G
COBBPN-8722-5GEA 0 GBP
COBBPN-8722-5G COBBPN-8722-1G
2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester
2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester
Formula: C₁₉H₃₁BN₄O₄
MW: 390.2848 g/mol
MDL Number: MFCD07781250
CAS Number: 940284-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR