2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PN-8713-250M
COBBPN-8713-250MEA 0 GBP
COBBPN-8713-250M
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₁₇BF₃NO₃
MDL Number: MFCD07781172
CAS Number: 1073354-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR