2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5923-1G PN-5923-250M
COBBPN-5923-1GEA 0 GBP
COBBPN-5923-1G COBBPN-5923-250M
2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester
2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester
Formula: C₁₃H₂₂BN₃O₂
MW: 263.1400 g/mol
MDL Number: MFCD12546556
CAS Number: 1218791-46-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR