1-(2-Bromophenyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OT-1253-1G
COBBOT-1253-1GEA 0 GBP
COBBOT-1253-1G
1-(2-Bromophenyl)piperidine
1-(2-Bromophenyl)piperidine
Formula: C₁₁H₁₄BrN
MW: 240.1396 g/mol
MDL Number: MFCD04112586
CAS Number: 156808-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR