9-Bromophenanthrene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OT-0490-25G OT-0490-100G OT-0490-5G
COBBOT-0490-25GEA 0 GBP
COBBOT-0490-25G COBBOT-0490-100G COBBOT-0490-5G
9-Bromophenanthrene
9-Bromophenanthrene
Formula: C₁₄H₉Br
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 62 °C
Density: 1.4093 g/cm³ (101 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001174
CAS Number: 573-17-1
EINECS: 209-351-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR