α,α,α-Trifluoro-p-cresol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Hydroxybenzotrifluoride, 4-(Trifluoromethyl)phenol

OS-7653-25G OS-7653-5G OS-7653-100G
COBBOS-7653-25GEA 0 GBP
COBBOS-7653-25G COBBOS-7653-5G COBBOS-7653-100G
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
Formula: C₇H₅F₃O
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 45…47 °C
Density: 1,3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002363
CAS Number: 402-45-9
EINECS: 206-945-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR