2-Bromo-5-formylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde

OS-7600-5G OS-7600-25G OS-7600-1G
COBBOS-7600-5GEA 0 GBP
COBBOS-7600-5G COBBOS-7600-25G COBBOS-7600-1G
2-Bromo-5-formylpyridine
2-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR