2,6-Dichloro-5-fluoronicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

OS-7532-5G OS-7532-100G OS-7532-25G
COBBOS-7532-5GEA 0 GBP
COBBOS-7532-5G COBBOS-7532-100G COBBOS-7532-25G
2,6-Dichloro-5-fluoronicotinic acid
2,6-Dichloro-5-fluoronicotinic acid
Formula: C₆H₂Cl₂FNO₂
MDL Number: MFCD00799517
CAS Number: 82671-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR