(±)-Nipecotic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OS-7493-25G OS-7493-100G
COBBOS-7493-25GEA 0 GBP
COBBOS-7493-25G COBBOS-7493-100G
(±)-Nipecotic acid
(±)-Nipecotic acid
Formula: C₆H₁₁NO₂
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR