2-Bromo-6-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OS-7438-25G OS-7438-100G
COBBOS-7438-25GEA 0 GBP
COBBOS-7438-25G COBBOS-7438-100G
2-Bromo-6-methylpyridine
2-Bromo-6-methylpyridine
Formula: C₆H₆BrN
MDL Number: MFCD00040743
CAS Number: 5315-25-3
EINECS: 226-173-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR