3-Bromo-2-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OS-7406-100G OS-7406-25G OS-7406-5G
COBBOS-7406-100GEA 0 GBP
COBBOS-7406-100G COBBOS-7406-25G COBBOS-7406-5G
3-Bromo-2-chloropyridine
3-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 55 °C
MDL Number: MFCD00234007
CAS Number: 52200-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR