7-Nitroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IN-0019-1G IN-0019-5G IN-0019-25G
COBBIN-0019-1GEA 0 GBP
COBBIN-0019-1G COBBIN-0019-5G COBBIN-0019-25G
7-Nitroindole
7-Nitroindole
Formula: C₈H₆N₂O₂
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR