3-Bromo-5-(trifluoromethyl)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride

HC-6321-5G HC-6321-25G HC-6321-100G
COBBHC-6321-5GEA 0 GBP
COBBHC-6321-5G COBBHC-6321-25G COBBHC-6321-100G
3-Bromo-5-(trifluoromethyl)aniline
3-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD00236205
CAS Number: 54962-75-3
EINECS: 259-412-6
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR