Ethyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

HC-6275-1G HC-6275-5G
COBBHC-6275-1GEA 0 GBP
COBBHC-6275-1G COBBHC-6275-5G
Ethyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate hydrochloride
Ethyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate hydrochloride
Formula: C₁₀H₁₀BrClN₂O₂
MDL Number: MFCD09801004
CAS Number: 957062-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR