6-Bromo-1,3-dichloroisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

HC-6021-5G HC-6021-1G
COBBHC-6021-5GEA 0 GBP
COBBHC-6021-5G COBBHC-6021-1G
6-Bromo-1,3-dichloroisoquinoline
6-Bromo-1,3-dichloroisoquinoline
Formula: C₉H₄BrCl₂N
MW: 276.9448 g/mol
MDL Number: MFCD09258639
CAS Number: 552331-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR